Liter Box
# de catalogue 470112-470
Fournisseur: SI Manufacturing
Spécifications
- Description:Liter Box
- Cat. no.:470112-470
- Supplier No.:77864-A
Spécifications
A propos de cet article
A perfect cubic decimeter box that contains exactly 1 liter. Can be used to demonstrate the metric relationships among linear, volume, one and mass measurements.
Customers Who Bought This Also Bought