Liquid Convection Apparatus
Catalog # 470148-912
Supplier: VWR International
Specifications
- Description:Liquid Convection Apparatus
- Length:81/4"
- Cat. no.:470148-912
- Width:6"
- Supplier No.:P20318
Specifications
About this item
Effective Method of Showing Normally Invisible Currents
A rectangular glass tube has an opening at the top in which to pour water and introduce dye, which is available separately. When a corner of the tube is heated, the current is visible in the movement of the dye.
Customers Who Bought This Also Bought