Leishmania donovani, Spleen Slide
# de catalogue 470181-892
Fournisseur: Avantor
Spécifications
- Description:Leishmania donovani, Spleen
- Cat. no.:470181-892
- Stain:Iron Hematoxylin
- Supplier No.:WARD470181-892
- Orientation:Section
Spécifications
A propos de cet article
Human Blood Parasite
- Identifying features clearly distinguishable
- Rigorous quality control standards
Shows typical Leishman-Donovan bodies.
Customers Who Bought This Also Bought