Color LEDs
Fournisseur: MEICO LAMP
Certificats
A propos de cet article
Every color of the rainbow available!
- Use as a source of constant wavelength in your diffraction and modern physics experiments
Each of the five LEDs is of a specific wavelength.
Customers Who Bought This Also Bought