Premium Mineral Collection
# de catalogue 470151-164
Fournisseur: Geoscience Industries
Spécifications
- Description:Large Size Mineral Collection
- No. of Samples:12
- Cat. no.:470151-164
- Supplier No.:1752-000
Spécifications
A propos de cet article
Premium introductory collection features 12 palm-sized specimens that depict the variety of colors and forms found in minerals. Each specimen is numbered and keyed to an ID sheet featuring additional data about each item. Includes an information booklet and a magnifier.
Specimens are approximately 2" x 3"
Customers Who Bought This Also Bought