Large Compass
# de catalogue 470102-466
Fournisseur: United Scientific Supplies
Spécifications
- Description:Large compass
- Cat. no.:470102-466
- Supplier No.:CPL090
Spécifications
A propos de cet article
Large compass that can easily be seen by the whole classroom.
- Large scale: 4x4"
- 360 degree scale
This magnetic compass is extra large (90mm diameter) and includes a bright color plastic case with a clear plastic top.
Customers Who Bought This Also Bought