Preserved Lancelet
Fournisseur: Avantor
Certificats
A propos de cet article
Formalin preserved lancelet. Available in vials or jars.
- Evolutionary biology
- Comparative anatomy
Amphioxus. This is a small, fishlike, primitive animal that possesses a notochord, but lacks a brain. Large, mature specimens.
Customers Who Bought This Also Bought