Lactose Tolerance: Human Evolution Kit
# de catalogue 470233-886
Fournisseur: Avantor
Spécifications
- Description:Lactose tolerance: Human evolution kit
- No. of Lab Groups:5
- Cat. no.:470233-886
Spécifications
A propos de cet article
Students research why one-third of Americans are lactose intolerant, but most other cultures have a much higher intolerance rate.
- For grades 9 - 12
- Complete instructions included
- Teacher-designed lab
- Students research where and why lactose intolerance is high
- Purpose for students: to learn how to make inferences based on research
In this lab designed by teacher Therese Passerini, students test milk with a glucose strip and introduce the enzyme lactase. After retesting, students notice that lactose in milk breaks down into galactose and glucose.
Students then research a world map to observe trends in lactose tolerance and make inferences on the inheritance and possible adaptive advantage of lactose tolerance in some populations. This kit is for grades 9 - 12.
Ordering information: This kit includes lactase, glucose test strips, map of the world, world map (color-coded), pedigrees, and complete instructions. Milk is not included. Required but not included: Test tubes, test tube racks, graduated cylinder.
Customers Who Bought This Also Bought