Student Grade Lab Pack Plastic Slide Set
# de catalogue 470152-874
Fournisseur: Avantor
Spécifications
- Description:Lab pack plastic slides set
- Cat. no.:470152-874
- Supplier No.:WARD470152-874
Spécifications
A propos de cet article
These blank microscope slides provide unsurpassed quality and value. Explore slidemaking with this starter pack.
- Hands-on learning
- High quality plastic
- Excellent clarity
Made of unbreakable plastic, these slides won't scratch or cut.
Delivery: Set Includes: 50 unbreakable slides (25 x 75 mm), 50 cover slips (22 mm square), 50 sheets of lens tissue.
Customers Who Bought This Also Bought