Laboratory Techniques Chart
# de catalogue 470008-196
Fournisseur: Denoyer-Geppert Science
Spécifications
- Description:Laboratory Techniques Chart
- Cat. no.:470008-196
- Supplier No.:2028-10
Spécifications
A propos de cet article
A quick reference guide and refresher, this chart is a great companion to our laboratory safety chart, 71093-00.
- Printed on tear-proof stock with eyelets for hanging
- Size: 35×44"
The clear and thorough illustrations answer questions on proper adjusting of burner flames, how to measure liquids, bend glass tubing, and 20 other techniques students need to master successfully.
Customers Who Bought This Also Bought