Laboratory Magnet Set
# de catalogue 470200-924
Fournisseur: United Scientific Supplies
Spécifications
- Description:Laboratory Magnet Set
- Cat. no.:470200-924
- Supplier No.:MSET4
Spécifications
A propos de cet article
A must for the Lab
- Set of four
The magnets in this set are permanent Alnico and include two bars, two horseshoes, and keepers. Sizes: bars 3"L x 5/8"W x 3/8"H; small horseshoe 1 "W x 1"H x 3/8"D; large horseshoe 1 3/8"W x 1 3/8"H x 3/4"D.
Customers Who Bought This Also Bought