Laboratory Engraver
Fournisseur: ROBERT BOSCH TOOL CORPORATION
Certificats
A propos de cet article
Protect your valuable microscopes, laboratory equipment, and apparatus with this easy-to-use engraver.It permanently marks on steel, plastic, copper, brass, wood, and glass. It handles like a pencil and has an easily replaceable carbide point.
The high speed, adjustable reciprocating motor delivers 7200 strokes per minute and is designed for trouble free, heavy duty use. Complete with cord and plug for operation on 110 V, ac.
Customers Who Bought This Also Bought