Laboratory Chow
# de catalogue 470105-576
Fournisseur: INDUSTRIAL SUPPORT INC.
Small Business Enterprise
Spécifications
- Description:Laboratory Chow
- Cat. no.:470105-576
- Supplier No.:21-1926
Spécifications
A propos de cet article
Use this complete diet for mice, gerbils, rats and hamsters.
The average monthly feed consumption for each animal is as follows: mouse - 150 g; gerbil - 200 g; rat - 450 g; and hamster - 300 g. Includes 5kg.
Customers Who Bought This Also Bought