ContactUsLinkComponent
Kyoto Kagaku® Pediatric I.V. Hand Trainer
Kyoto Kagaku® Pediatric I.V. Hand Trainer
# de catalogue 470126-220
PEDIATRIC IV HAND SIM
Kyoto Kagaku® Pediatric I.V. Hand Trainer
# de catalogue 470126-220
Spécifications
  • Description:
    Pediatric IV Hand
  • Cat. no.:
    470126-220
  • Supplier No.:
    M95
Spécifications
A propos de cet article
Practice Pediatric Injections
  • Three accessible puncture sites
  • Swivel stand allows for proper positioning
Higher skills are required to perform IV procedures safely on the hands of infants who have soft veins that are difficult to find. This simulator is an effective tool providing hands-on training as a life-like model. The back of the hand and fingers are flexible and can be gripped or pulled as in real procedures. Two puncture sites can be found on the back of the hand, and the needle fixing procedure can also be practiced.
Delivery: For distribution within North America only.
Customers Who Bought This Also Bought