Kidney, Human Slide
# de catalogue 470176-972
Fournisseur: Avantor
Spécifications
- Description:Kidney, Human Slide
- Cat. no.:470176-972
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470176-972
- Orientation:Section
Spécifications
A propos de cet article
Illustrates kidney form
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. Cortex and medulla. Shows renal corpuscle, tubule, and collecting duct
Customers Who Bought This Also Bought