ContactUsLinkComponent
Kidney, Human Slide
Kidney, Human Slide
# de catalogue 470176-972
Fournisseur:  Avantor
KIDNEY (HUMAN) CS WARDS ITEM
Kidney, Human Slide
# de catalogue 470176-972
Fournisseur:  Avantor
Spécifications
  • Description:
    Kidney, Human Slide
  • Cat. no.:
    470176-972
  • Stain:
    Hematoxylin and Eosin
  • Supplier No.:
    WARD470176-972
  • Orientation:
    Section
Spécifications
A propos de cet article
Illustrates kidney form
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Human. Cortex and medulla. Shows renal corpuscle, tubule, and collecting duct
Customers Who Bought This Also Bought