Kid Inventor Basic Creative Electronics Kit
# de catalogue 470100-800
Fournisseur: SHINET CORP
Spécifications
- Description:Kit, Kid Inventor, Basic 120 Projects, Creative Electronics
- Cat. no.:470100-800
- Supplier No.:K-120
Spécifications
A propos de cet article
Using the materials in this kit young inventors can build over 120 projects, including a magnetic fan, a sound-activated musical doorbell, and a fingertip flashbulb.
Each project is constructed using components with snap connections that make assembly easy and secure, so there is no need for soldering. Comes with manual.
Ordering information: Four AA batteries required, but not included.
Customers Who Bought This Also Bought