Karyotyping Simulation Kit
# de catalogue 470024-092
Fournisseur: Science First
Spécifications
- Description:Karyotyping Simulation Kit
- Cat. no.:470024-092
- Supplier No.:6900
Spécifications
A propos de cet article
Illustrate the Relationship Between Karyotyping and Genetic Diseases
Lead students on the classification of a normal male, an individual with Kleinfelter’s Syndrome, and one with Turner’s Syndrome using this hands-on activity. After cutting out the chromosomes, students use the provided karyotyping trays to facilitate sorting. Ordering chromosomes which have been frozen at the metaphase state, students identify the different genetic diseases.
Customers Who Bought This Also Bought