Rescue Critters® Emily K-9 Positioning Manikin
# de catalogue 470029-832
Fournisseur: THALES & CO.
Spécifications
- Description:K-9 Positioning Manikin
- Cat. no.:470029-832
- Supplier No.:300
Spécifications
A propos de cet article
Featuring Realistic Rotation And Range Of Motion
- Natural articulation and range of motion
- Can be adjusted to correct positions for surgery and x-ray
- Made to order
Students learn how to position dogs for abdominal surgery, x-ray, spay and neuter, advanced spinal stabilization, spinal recumbency and more. It has natural movements and offers resistance; allowing you to safely practice techniques before working on live animals. Fully articulated, Emily has shoulders and hip joints that rotate realistically, with 90° flexibility. Also with 90° range of motion are the knees, elbows, and carpal and hock joints. This baby comes complete with carrying case, kneeling pad and brush.
Ordering information: This item is made to order. Returns cannot be accepted.
Customers Who Bought This Also Bought