Jumper Wire Set
# de catalogue 470110-328
Fournisseur: GLOBAL SPECIALTIES SE
Spécifications
- Description:Jumper Wire Set
- Cat. no.:470110-328
- Supplier No.:111-0044
Spécifications
A propos de cet article
The pre-formed Jumper Wire Set includes 25 wires in 14 color-coded lengths.
- Twenty-Five Pieces
- Fourteen Color-Coded Lengths
- Convenient Plastic Case
- For Making Connections on Breadboard Circuits
- Set Keeps Circuits Organized
The wires can be used to make connections on breadboard circuits.
It's an essential timesaver that keeps circuits clear and organized.
Customers Who Bought This Also Bought