ContactUsLinkComponent
Jumbo LED 10 mm
Jumbo LED 10 mm
# de catalogue 470188-768
Fournisseur:  Avantor
LED JUMBO YELLOW 10 MM
Jumbo LED 10 mm
# de catalogue 470188-768
Fournisseur:  Avantor
Spécifications
  • Description:
    LED, Jumbo, Yellow, 10 mm
  • Cat. no.:
    470188-768
  • Supplier No.:
    4646903
Spécifications
A propos de cet article
Bright and colorful LED lamps for any simple circuit experiment.
A variety of colors are available.
Customers Who Bought This Also Bought