Jiffy® Pots
# de catalogue 470150-548
Fournisseur: PLANTATION PRODUCTS, INC.
Spécifications
- Description:Jiffy Pots, 8 cm x 8 cm, Jiffy Products of America
- Cat. no.:470150-548
- Supplier No.:JP310
Spécifications
A propos de cet article
Plant roots can grow with ease through these pot walls, allowing you to replant without removing the pot. Because transfer is avoided, the plant may develop more quickly than usual.
Customers Who Bought This Also Bought