Iron Meteorite (Type IAB)
# de catalogue 470155-354
Fournisseur: ADAM MOORE MO
Spécifications
- Description:Iron Meteorite (Type IAB)
- Supplier No.:495695
Spécifications
A propos de cet article
Classic nickel-iron meteorite with excellent surface quality from a famous find in Campo del Cielo. Average size: 1" x 2 1/2". Weight: 320–380g. Argentina.
Customers Who Bought This Also Bought