Ion Exchange Chromatography Activity Kit
# de catalogue 470200-750
Fournisseur: G-Biosciences
Spécifications
- Cat. no.:470200-750
- Supplier No.:BE-415
Spécifications
A propos de cet article
Illustrate Column Chromatography Fundamentals
Most biological molecules have a net charge within a pH range of 2 to 10. When the pH is altered, the net charge on molecules can change drastically. In this experiment, proteins are absorbed onto a charged solid support ion-exchange column and subsequently separated during elution using a two-step salt gradient. Students learn basic principals of column chromatography and practice techniques of packing a column, eluting and collecting column fractions, and performing analysis of column fractions through spectrophotometry. Includes materials for six setups.
Customers Who Bought This Also Bought