ContactUsLinkComponent
Ward's® Investigating Antibiotic Resistance Through Conjugation Lab Activity
Ward's® Investigating Antibiotic Resistance Through Conjugation Lab Activity
# de catalogue 470193-394
Fournisseur:  MICROLOGY INC
WARD'S Investig Antibiotic Resistance
Ward's® Investigating Antibiotic Resistance Through Conjugation Lab Activity
# de catalogue 470193-394
Fournisseur:  MICROLOGY INC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Ward's® Investigating Antibiotic Resistance
  • No. of Lab Groups:
    6
  • Cat. no.:
    470193-394
  • Supplier No.:
    K30IA
Spécifications
A propos de cet article
Identify Host and Donor Bacteria in Gene Transfer Activity
  • Make concrete, the abstract concept of conjugation for your students.
  • Introducte the concept of antibiotic resistance with this hands-on activity
  • Easygel media saves you time, no need to melt your media
Conjugation, one method of transferring antibiotic-resistance, or R- factors, to bacteria is explored in this lab.
Students transfer the genes for antibiotic resistance between different strains of bacteria, then determine which bacteria are the hosts and which are the donors, and whether chromosomal or plasmid DNA was transferred.
For even greater convenience, the lab activity features Easygel, a premeasured, sterilized media that requires no autoclave or hot waterbath. The activity contains enough materials for six setups, a teacher’s guide, and student copymaster.
Ordering information: This item contains live or perishable materials. At checkout, please provide the date you wish the complete kit to arrive.
Delivery: Kit will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Customers Who Bought This Also Bought