Ward's® Introductory Rock Collection
# de catalogue 470025-228
Fournisseur: Avantor
Spécifications
- Description:Introductory Rock Collection
- No. of Samples:12
- Cat. no.:470025-228
- Supplier No.:WARD470025-228
Spécifications
A propos de cet article
Ideal for Beginning Earth Science Students
- Cut by WARD's Geologists
- Highest Quality Specimens
- Great for Introducing Students to Rock types
Four typical rocks from each of the three rock groups — igneous, sedimentary, and metamorphic — comprise our most basic collection.
The set is housed in a compartmented collection box and includes an identification list.
Contents:
12 numbered samples, 1" x 2"
Igneous: basalt, gabbro, granite, rhyolite
Sedimentary: conglomerate, limestone, sandstone, shale
Metamorphic: gneiss, marble, quartzite, schist
Customers Who Bought This Also Bought