Ward's® Introductory Mineral and Rock Collection
# de catalogue 470025-166
Fournisseur: Avantor
Spécifications
- Description:WARD'S introductory mineral and rock collection
- Supplier No.:WARD470025-166
Spécifications
A propos de cet article
Examples of the most basic rocks and rock-forming minerals.
- Sixteen 1×2" numbered samples
- Housed in a compartmented collection box
- Identification key included
Give students a clear introduction to rocks and rock-forming minerals; the specimens in the introductory set are easy to differentiate from one another and clearly show the traits of each type.
Minerals: Biotite, feldspar, quartz, calcite
Igneous: Basalt, granite, rhyolite, gabbro
Sedimentary: Conglomerate, limestone, sandstone, shale
Metamorphic: Gneiss, marble, quartzite, schist
Customers Who Bought This Also Bought