Introductory Earth Science Collection
# de catalogue 470112-322
Fournisseur: SCOTT RESOURCES 2, LLC SE
Spécifications
- Description:Introductory Earth Science Collection
- No. of Samples:75
- Cat. no.:470112-322
- Supplier No.:2175
Spécifications
A propos de cet article
Introduce students to a vast variety of rocks and minerals
- Introduces Students to all types of rocks
- Study guide included
Students can observe, compare and test these 75 mineral and rock specimens contained in this comprehensive collection. Each sample measures 1" x 1-1/2".
Includes igneous rocks, sedimentary rocks, metamorphic rocks, rock forming minerals, hardness scale samples (diamond not included) and specimens illustrating physical properties. Comes with a streak plate, magnifier, key sheet and an English-Spanish study guide.
Customers Who Bought This Also Bought