Ward's® Crystal Form Collection
# de catalogue 470015-404
Fournisseur: Avantor
Spécifications
- Description:Introductory Crystal Form Collection
- No. of Samples:6
- Cat. no.:470015-404
- Supplier No.:WARD470015-404
Spécifications
A propos de cet article
Basic Set of Natural Crystals
- Hand Selected by WARD's Geologists
- Teach Students the Basic Crystal Forms
The six main crystal systems are represented in this beginning geology student’s introduction to crystal form. The set is housed in a compartmented collection box, and an identification list is included.
Contents:6 numbered samples, 1" x 1"
Isometric Pyrite
Hexagonal Quartz
Tetragonal Zircon
Orthorhombic Danburite
Monoclinic Orthoclase
Triclinic Microcline
Customers Who Bought This Also Bought