Introduction to Marine Biology Microslide
Fournisseur: SUPERTEK SCIENTIFIC LLC
Certificats
A propos de cet article
Explore Sea life Microscopically
- Arrows help students locate important features
- Mounted in a clear plastic holder that protects it on both sides
Microslides consist of a strip of film with photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides. A detailed text folder is included. Viewers are available separately.
Ordering information: Set includes Phytoplankton (50X), Zooplankton (12 1/2X), Sponge spicules (30X), Coral polyps (6X), Jellyfish tentacle (cs, 100X), Starfish (1X), Featherworms ( 1/2X), and Shark scales (13X).
Customers Who Bought This Also Bought