Instant Ear Thermometer
# de catalogue 470018-918
Fournisseur: GF HEALTH PRODUCTS INC. TE
Spécifications
- Description:Instant Ear Thermometer
- Cat. no.:470018-918
- Supplier No.:2215
Spécifications
A propos de cet article
This thermometer features one-touch operation and avoids the hassle of oral thermometers.
- Accurate Readout in One Second
- Fever Alarm
The user-friendly instrument yields accurate results within one second of being placed in the patient’s ear. A fever alarm sounds for temperatures above 100.4°F. The thermometer's display indicates measurements in either Celsius or Fahrenheit, and the Nite-Glo display can be seen in any lighting. The thermometer has a range of 93.2°F to 109.4°F and includes a storage case, lithium battery, and guidebook.
Customers Who Bought This Also Bought