Insect Identification Poster
# de catalogue 470007-218
Fournisseur: FEENIXX PUBLISHING, INC.
Spécifications
- Length:92 cm
- Width:61 cm
- Description:Insect Poster
- Cat. no.:470007-218
- Finish:Laminated
- Supplier No.:A222L
Spécifications
A propos de cet article
A Unique Branching System Classifies By Insect Order
- The branching organization system helps students with quick identification
A brief description and photo of the most common species accompanies each insect order featured. Brief description of insect anatomy and other arthropods, like spiders and crustaceans, are also included.
Customers Who Bought This Also Bought