Inflatable Solar System
# de catalogue 470149-810
Fournisseur: Learning Resources
Spécifications
- Description:Inflatable Solar System Model
- Cat. no.:470149-810
- Supplier No.:LER2434
Spécifications
A propos de cet article
Hang the Whole Solar System From Your Ceiling
You can teach size and distance relationships of planets, the moon, and the sun with this inflatable set. Students can learn about the rotation, revolution, and orbit through class activities or demonstrations. Each object can be deflated for easy storage. The kit comes with a 36" dia. Sun, proportionate planets and moon ranging in size from 8–22", a foot pump, teacher’s guide, and hooks for easy hanging.
Customers Who Bought This Also Bought