Indoor/Outdoor Thermometer and Hygrometer
# de catalogue 470104-560
Fournisseur: Taylor Precision Products
Spécifications
- Description:Indoor/outdoor thermometer with hygrometer, -45 to 120 F, red liquid filled
- Cat. no.:470104-560
- Supplier No.:5329
Spécifications
A propos de cet article
This combination unit indicates the inside and outside temperatures and the indoor humidity.
- Outside thermometer is connected to a remote sensing bulb by armored tubing
- Contains red fluid
Measures 24x6 cm
Ordering information: Includes mounting hardware.
Customers Who Bought This Also Bought