Immersion Oil Cleaner
# de catalogue 470178-434
Fournisseur: Ward's Science
Numéro CAS:
Spécifications
- Description:Immersion Oil Cleaner, Laboratory
- Size:15 mL
- Cat. no.:470178-434
- Supplier No.:IX0231-15
Spécifications
A propos de cet article
Use this solution to clean immersion oil off microscope objectives.
Customers Who Bought This Also Bought