Illustrated Dissection Guides
# de catalogue 470146-760
Fournisseur: Ward's Science
Spécifications
- Description:Rabbit Guide
- Cat. no.:470146-760
- No. of Pages:25
- Length:22 cm
- Width:14 cm
- Supplier No.:420.41341
Spécifications
A propos de cet article
These economical and concise guides contain the right amount of information for reference during dissection.
- Basic Facts on Specimens
- Convenient Size
- Paperback
- Color Illustrations
- Step-by-Step Instructions
Each booklet provides basic facts on specimens, step-by-step dissection instructions, and clear illustrations to guide students as they dissect their own specimens.
The paperback guides are small, so students can have them whenever they're needed.
Customers Who Bought This Also Bought