Ileum, Human Slide
# de catalogue 470176-994
Fournisseur: Avantor
Spécifications
- Description:Ileum, Human
- Cat. no.:470176-994
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470176-994
- Orientation:Section
Spécifications
A propos de cet article
Final Section of Small Intestine
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. Its function is to absorb vitamin B12, excess bile salts, and other digestive products.
Customers Who Bought This Also Bought