Igneous Rocks Classroom Project
# de catalogue 470201-208
Fournisseur: SCOTT RESOURCES 2, LLC SE
Spécifications
- Description:Igneous rock chart
- Cat. no.:470201-208
- Supplier No.:2506
Spécifications
A propos de cet article
The purpose of this activity is to become acquainted with the origin, texture, composition, and interrelationships of igneous rocks. This is accomplished by providing a key to the systematic arrangement of the igneous rocks and graphically displaying these properties.
Customers Who Bought This Also Bought