Hypothalamus Slide
# de catalogue 470182-726
Fournisseur: Avantor
Spécifications
- Description:Hypothalamus
- Cat. no.:470182-726
- Stain:Cresyl violet
- Supplier No.:WARD470182-726
- Orientation:Cross section
Spécifications
A propos de cet article
Multifunctional Region of the Brain
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Shows third ventricle and optic nerve.
Customers Who Bought This Also Bought