Human Skin, Composite Slide
# de catalogue 470183-070
Fournisseur: Avantor
Spécifications
- Description:Human Skin, Composite Slide
- Cat. no.:470183-070
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470183-070
- Orientation:Section
Spécifications
A propos de cet article
Study human epidermis
- Identifying features clearly distinguishable
- Rigorous quality control standards
Pigmented and unpigmented adult skin with all layers, epidermis, dermis, and subcutaneous tissue, for easy comparison.
Customers Who Bought This Also Bought