Eisco® Human Male Reproductive System Model
# de catalogue 470111-068
Fournisseur: VWR International
Spécifications
- Description:Male Pelvic
- Height:8 cm
- Cat. no.:470111-068
- Length:34 cm
- Width:36 cm
- Supplier No.:AM0118
- No. of Parts:1
Spécifications
A propos de cet article
Rectum, bladder with prostate and testicular ducts are clearly shown on this life-size models that offers views of both the external and internal male genital organs. Model is numbered, mounted on base, and includes a key card.
Customers Who Bought This Also Bought