ContactUsLinkComponent
Human Genetics Slide Set
Human Genetics Slide Set
# de catalogue 470183-612
Fournisseur:  Avantor
HUMAN GENETICS SET SET/6
Human Genetics Slide Set
# de catalogue 470183-612
Fournisseur:  Avantor
Spécifications
  • No. of Slides:
    6
  • Description:
    Human Genetics Slide Set
  • Cat. no.:
    470183-612
  • Type:
    Animal
  • Supplier No.:
    WARD470183-612
Spécifications
A propos de cet article
Illustrates the Complexity of the Human Chromosome
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
All are smears unless otherwise noted.
Ordering information: Set includes Chromosomes, human male 46 XY; Chromosomes, human female 46 XX; Peripheral blood, human male; Peripheral blood, human female; Squamous epithelium
comparison, human male/female buccal scrapings; and Barr bodies in neutrophils (cs).
Customers Who Bought This Also Bought