Human Genetics Slide Set
# de catalogue 470183-612
Fournisseur: Avantor
Spécifications
- No. of Slides:6
- Description:Human Genetics Slide Set
- Cat. no.:470183-612
- Type:Animal
- Supplier No.:WARD470183-612
Spécifications
A propos de cet article
Illustrates the Complexity of the Human Chromosome
- Identifying features clearly distinguishable
- Rigorous quality control standards
All are smears unless otherwise noted.
Ordering information: Set includes Chromosomes, human male 46 XY; Chromosomes, human female 46 XX; Peripheral blood, human male; Peripheral blood, human female; Squamous epithelium
comparison, human male/female buccal scrapings; and Barr bodies in neutrophils (cs).
comparison, human male/female buccal scrapings; and Barr bodies in neutrophils (cs).
Customers Who Bought This Also Bought