Eisco® Female Pelvic Model
# de catalogue 470111-066
Fournisseur: VWR International
Spécifications
- Description:Female Pelvic
- Height:8 cm
- Cat. no.:470111-066
- Length:34 cm
- Width:36 cm
- Supplier No.:AM0116A
- No. of Parts:1
Spécifications
A propos de cet article
This life size model highlights all female genital organs, with both the bladder and rectum fully exposed.
- Structures are numbered
Mounted on a base, the model includes a key card.
Customers Who Bought This Also Bought