A propos de cet article
Detailed images of human blood and cells.
- Arrows help students locate important features
- Mounted in a clear plastic holder that protects it on both sides
- Set of 15 includes detailed lesson plan on the topic under study
This set of microslides allows students to explore the many cells that make up their blood. They will see that there's more to blood than meets the eye.
Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides. A detailed text folder is included. Viewers are available separately.
Contents:
Human blood smear (st, 900X)
Red corpuscles (em, 3,000X)
White corpuscles/leukocytes (sm, st, 1,400X)
Phagocytosis (em, 25,000X, 3,500X)
Blood type B (wem, 100X)
Fibrin, washed clot (400X)
Sickle cells (sm, st, 100X)
Infected blood (sm, st, 800X)
Box of 15 sets also Includes:
Background subject information
Reproducible student worksheets with answer keys+T30