Human Blood Slide
# de catalogue 470177-632
Fournisseur: Avantor
Spécifications
- Description:Human Blood, smear
- Cat. no.:470177-632
- Stain:Wright's
- Supplier No.:WARD470177-632
- Orientation:Smear
Spécifications
A propos de cet article
Smear, Wright's stain
- All identifying features are clearly distinguishable
- Rigorous quality control standards
Shows the three major functional classes: red cells, white cells, and platelets.
Customers Who Bought This Also Bought