Large Wave form helix
Fournisseur: VWR International
Certificats
A propos de cet article
The Large Wave form helix easily demonstrates wave phenomena.
- Students Research Behavior of Pulses, Traveling Waves, and Standing Waves
- Stand Makes for Easy Storage
- For Demonstrating Wave Phenomena
With this demonstration, students can conceptualize the behavior of pulses, traveling waves, and standing waves.
With tangling woes also an issue, the featured stand provides easy deployment, retrieval, and storage.
Size of slinky spring (without stand) is 75mm (±2mm) diameter. Coil close length is 110mm (±5mm).
Customers Who Bought This Also Bought