ContactUsLinkComponent
Horse Ovary
Horse Ovary
# de catalogue 470001-392
Fournisseur:  Avantor
HORSE OVARY PRESERVED JAR/EACH
Horse Ovary
# de catalogue 470001-392
Fournisseur:  Avantor
Spécifications
  • Description:
    Horse Ovary, Preserved
  • Cat. no.:
    470001-392
  • Supplier No.:
    WARD470001-392
Spécifications
A propos de cet article
This large specimen can be used as an alternative to a pig or sheep ovary. Dissection of the horse ovary illustrates the functions of this major organ of the female mammalian reproductive system.
Customers Who Bought This Also Bought