ContactUsLinkComponent
Horse Digestive Tract
Horse Digestive Tract
# de catalogue 470000-448
Fournisseur:  Avantor
HORSE DIGESTIVE TRACT
Horse Digestive Tract
# de catalogue 470000-448
Fournisseur:  Avantor
Spécifications
  • Description:
    Horse Digestive Tract, Preserved
  • Cat. no.:
    470000-448
  • Supplier No.:
    WARD470000-448
Spécifications
A propos de cet article
Formalin Preserved
Give students an in-depth anatomy lesson with this specimen which includes a complete alimentary canal: esophagus, stomach, small and large intestines, and colon. The horse digestive tract is a much larger alternative to the sheep or goat for clear illustrations. Note: Additional freight charges may apply.
Customers Who Bought This Also Bought