Ward's® Hornblende (Cleavage)
Fournisseur: Avantor
Certificats
A propos de cet article
Amphibole group; dark greenish black cleavages, somewhat columnar.
Ordering information: WARD’S offers different specimen sizes to meet your various needs. Student specimens, hand specimens, and bulk packs are available.
Customers Who Bought This Also Bought