Spécifications
- Description:Tube Shield for Horizontal Spin Centrifuge
- Cat. no.:470144-636
- Supplier No.:C800-02
Spécifications
A propos de cet article
With speeds up to 5500 RMP and horizontal spin rotor this centrifuge is a great instrument for a busy laboratory.
- Variable Speed Range 500-5,500 RPM
- All digital front panel with 3 pre-programmed options
- Custom settings
Featuring variable speed range of 500-5,500 RMP, to ensure a clean and complete separation. Quiet and dependable, with a maintenance free motor, stainless steel tube shields it can be used to process blood, urine, fecal and other specimens. The all digital front panel with 3 pre-programmed options, 3 Minutes at 4,000G, 5 minutes at 3,200G and 10 minutes at 2,000G, provides accurate results every time. Custom settings are easy and intuitive with all digital panel and oversized display.
Features and benefits include Zero RPM Safety Latching System provides lid from being opened while rotor is spinning. Quiet running, only 50 decibels at 3500 RPM. 99 minute digital timer with one second increments. Remembers last setting for quick repeat runs. Adjustable acceleration and deceleration. Stainless steel rotor chamber for easy cleaning. Suction cup rubber feet eliminate vibration. Available in 110/220V 50/60HZ.