DuraHoop™ Hula Hoop, GOPHER SPORT
# de catalogue 470146-228
Fournisseur: GOPHER SPORT
Spécifications
- Description:DuraHoop™ Hula Hoop
- Cat. no.:470146-228
- Supplier No.:10-224
- Diameter:24"
Spécifications
A propos de cet article
Hoop that is safer and more durable with fused, staple-free ends.
- Featuring a 2.3 mm thick wall
- Rigid design makes it kink-proof, even when stepped on
There is much less of a risk of the hoops coming apart and students getting injured when using these.
The thickness makes the hoop kink-proof and able to hold up to long-term institutional play.
Customers Who Bought This Also Bought